Product Details

SNP ID
rs2561482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:103262097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTTTGTATATTTTTGGGAAAG[C/G]CACTCCCTTTTTTGGGTTCCTAGTG
Phenotype
MIM: 616608
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C5orf30 PubMed Links

Gene Details

Gene
C5orf30
Gene Name
chromosome 5 open reading frame 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316968.1 Intron NP_001303897.1
NM_001316969.1 Intron NP_001303898.1
NM_033211.3 Intron NP_149988.1

View Full Product Details