Product Details

SNP ID
rs2562740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128492078 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGTGGTACAATCAAGCTGACTGC[A/G]GCCTTGATCTCCTGGGCTCAAGTGA
Phenotype
MIM: 607846
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL2B PubMed Links

Gene Details

Gene
METTL2B
Gene Name
methyltransferase like 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018396.2 Intron NP_060866.2

View Full Product Details