Product Details

SNP ID
rs2638539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118813772 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATCTATTCTGACCAAAAACAGTA[C/T]ATAATTTCCTTGGTTTTAAAGACAA
Phenotype
MIM: 610098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCM9 PubMed Links

Gene Details

Gene
MCM9
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017696.2 4499 UTR 3 NP_060166.2
NM_153255.4 4499 Intron NP_694987.1

View Full Product Details