Product Details
- SNP ID
-
rs2640713
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:10367771 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACTAAGGACTTGAAGCTTTTTTTT[A/T]ATTAAAAAAACCACTATTGTTTATT
- Phenotype
-
MIM: 613297
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MARCH6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113081329] are located under a probe and SNP(s) [rs74661932] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MARCH6
- Gene Name
- membrane associated ring-CH-type finger 6
View Full Product Details