Product Details
- SNP ID
-
rs2663920
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:81104063 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTCCAGATGGGAAAATAGAGGC[G/A]CAGAGAGGAGAACAAAAACTGCTCA
- Phenotype
-
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CFAP161
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs57360728,rs61410377] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CFAP161
- Gene Name
- cilia and flagella associated protein 161
View Full Product Details