Product Details

SNP ID
rs2682500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10415448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAAACTGACCAACATAACATGCA[A/G]CTTTTAGGAAGCCTGAATGCCACAA
Phenotype
MIM: 602892 MIM: 602893
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLRC3 PubMed Links

Gene Details

Gene
KLRC3
Gene Name
killer cell lectin like receptor C3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002261.2 Intron NP_002252.2
NM_007333.2 Intron NP_031359.2
Gene
KLRC4
Gene Name
killer cell lectin like receptor C4
There are no transcripts associated with this gene.

Gene
KLRC4-KLRK1
Gene Name
KLRC4-KLRK1 readthrough
There are no transcripts associated with this gene.

View Full Product Details