Product Details
- SNP ID
-
rs2254303
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:43308652 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTACCAGCCGCTGCAGGTGAAAAGG[G/A]TGTCTGGGACTACACTGTGGCTAGC
- Phenotype
-
MIM: 604995
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRIP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12193537] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRIP3
- Gene Name
- cysteine rich protein 3
- Gene
- SLC22A7
- Gene Name
- solute carrier family 22 member 7
There are no transcripts associated with this gene.
View Full Product Details