Product Details
- SNP ID
-
rs2705329
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:34232550 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTGAAGTTTCTGAAAACAAATTA[A/G]AAGACTGTTGGCTTGGCTAATCTCG
- Phenotype
-
MIM: 616245
MIM: 604878
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EMC4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116076857] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EMC4
- Gene Name
- ER membrane protein complex subunit 4
There are no transcripts associated with this gene.
- Gene
- SLC12A6
- Gene Name
- solute carrier family 12 member 6
View Full Product Details