Product Details
- SNP ID
-
rs2706226
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:54632759 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAACAAGTTCAATTTAGCTGATG[G/T]GGGTGCGACATAAAAGAGAGTCATG
- Phenotype
-
MIM: 607360
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LACRT
PubMed Links
Gene Details
- Gene
- LACRT
- Gene Name
- lacritin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033277.1 |
|
Intron |
|
|
NP_150593.1 |
View Full Product Details