Product Details
- SNP ID
-
rs2677893
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:2572334 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATCATGTGTATCCTAGGAGACAAA[T/C]TTTTTAGCAGATGTGAAGGGAGTGA
- Phenotype
-
MIM: 607272
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
METTL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76051747,rs9953982] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- METTL4
- Gene Name
- methyltransferase like 4
- Gene
- NDC80
- Gene Name
- NDC80, kinetochore complex component
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006101.2 |
|
Intron |
|
|
NP_006092.1 |
View Full Product Details