Product Details

SNP ID
rs2698192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61030850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAGGTAGATTCATGGTTGCCA[A/C]AGATGGGTGGAGGGGTGGGCAGGAA
Phenotype
MIM: 601789
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PEX13 PubMed Links

Gene Details

Gene
PEX13
Gene Name
peroxisomal biogenesis factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002618.3 Intron NP_002609.1

View Full Product Details