Product Details

SNP ID
rs2742111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10172555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCAGATATATTCTCTTTTGATT[A/T]TCACTGTTTTATCTGGCAATGGATC
Phenotype
MIM: 602601
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OLR1 PubMed Links

Gene Details

Gene
OLR1
Gene Name
oxidized low density lipoprotein receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172632.1 Intron NP_001166103.1
NM_001172633.1 Intron NP_001166104.1
NM_002543.3 Intron NP_002534.1
Gene
TMEM52B
Gene Name
transmembrane protein 52B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079815.1 Intron NP_001073283.1
NM_153022.3 Intron NP_694567.1
XM_006719029.2 Intron XP_006719092.1

View Full Product Details