Product Details

SNP ID
rs2255594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:113080578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATAATTTTTTCATCTAATCACT[A/G]TGGTCAATTTTGTGTTATTTAAATG
Phenotype
MIM: 605188
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR85 PubMed Links

Gene Details

Gene
GPR85
Gene Name
G protein-coupled receptor 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146265.1 4768 UTR 3 NP_001139737.1
NM_001146266.1 4768 UTR 3 NP_001139738.1
NM_001146267.1 4768 UTR 3 NP_001139739.1
NM_018970.6 4768 UTR 3 NP_061843.3
XM_005250451.2 4768 Intron XP_005250508.1
XM_017012352.1 4768 Intron XP_016867841.1

View Full Product Details