Product Details

SNP ID
rs2256652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100585733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGGGGTTCTCAAAGATGGGAGA[A/G]GATGGCTGCAGGTTTAGGGCAATCA
Phenotype
MIM: 609097
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO24 PubMed Links

Gene Details

Gene
FBXO24
Gene Name
F-box protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163499.1 Intron NP_001156971.1
NM_012172.4 Intron NP_036304.2
NM_033506.2 Intron NP_277041.1
XM_005250259.3 Intron XP_005250316.1
XM_011516022.1 Intron XP_011514324.1
XM_011516023.2 Intron XP_011514325.1
XM_017011961.1 Intron XP_016867450.1
Gene
LRCH4
Gene Name
leucine rich repeats and calponin homology domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289934.1 Intron NP_001276863.1
NM_002319.4 Intron NP_002310.2
Gene
PCOLCE-AS1
Gene Name
PCOLCE antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details