Product Details

SNP ID
rs2735185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141637636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGCGGCGAAGGGGGAGGGGA[G/T]GAATCAAGGGATGAGCGCCGGAAGG
Phenotype
MIM: 605166 MIM: 611213
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCHSD1 PubMed Links

Gene Details

Gene
FCHSD1
Gene Name
FCH and double SH3 domains 1
There are no transcripts associated with this gene.

Gene
HDAC3
Gene Name
histone deacetylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003883.3 691 Intron NP_003874.2
XM_011537697.2 691 Intron XP_011535999.1
Gene
RELL2
Gene Name
RELT like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130029.1 691 UTR 5 NP_001123501.1
NM_173828.4 691 UTR 5 NP_776189.3
XM_005268414.3 691 UTR 5 XP_005268471.1
XM_011537624.1 691 UTR 5 XP_011535926.1
XM_011537625.2 691 UTR 5 XP_011535927.1

View Full Product Details