Product Details

SNP ID
rs2735606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8140656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGGCTGGAACCCCCGGGTCCTG[C/G]TTGGGCCTCAGGCTCTCCTCCCATC
Phenotype
MIM: 602260
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6883 PubMed Links

Gene Details

Gene
MIR6883
Gene Name
microRNA 6883
There are no transcripts associated with this gene.

Gene
PER1
Gene Name
period circadian clock 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002616.2 4343 UTR 3 NP_002607.2
XM_005256689.1 4343 UTR 3 XP_005256746.1
XM_005256690.1 4343 UTR 3 XP_005256747.1

View Full Product Details