Product Details

SNP ID
rs2759328
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:173907567 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGATGGGAGAAAGTTGGCTTCAA[C/T]CCACAGATGGGAATTCAGTTTGGAT
Phenotype
MIM: 107300
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINC1 PubMed Links

Gene Details

Gene
SERPINC1
Gene Name
serpin family C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000488.3 Intron NP_000479.1
XM_005245198.3 Intron XP_005245255.1

View Full Product Details