Product Details

SNP ID
rs45567037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44620212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATTCATGGCGCTGCCCAAGAA[C/T]GGAAGGGCATCTTGCTAGAAGTCCC
Phenotype
MIM: 608958 MIM: 604932
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADA PubMed Links

Gene Details

Gene
ADA
Gene Name
adenosine deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000022.3 Intron NP_000013.2
NM_001322050.1 Intron NP_001308979.1
NM_001322051.1 Intron NP_001308980.1
Gene
PKIG
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor gamma
There are no transcripts associated with this gene.

View Full Product Details