Product Details

SNP ID
rs3735644
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:127582103 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCTGGACTGAAGTGCAAGATAGTCA[G/A]TATGGCTGTGAGAGTCTGCTGGCGG
Phenotype
MIM: 103188 MIM: 607418
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ARF5 PubMed Links

Gene Details

Gene
ARF5
Gene Name
ADP ribosylation factor 5
There are no transcripts associated with this gene.

Gene
GCC1
Gene Name
GRIP and coiled-coil domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024523.5 2657 Silent Mutation CTG,TTG L747L NP_078799.2

View Full Product Details