Product Details

SNP ID
rs2234428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23557571 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCCCCCTTCAGCTTGGGAAAGCG[A/G]AAGCGTGGCAGGGGCCGGACCTGGG
Phenotype
MIM: 612799
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EARS2 PubMed Links

Gene Details

Gene
EARS2
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083614.1 37 Intron NP_001077083.1
NM_001308211.1 37 Intron NP_001295140.1
XM_011545738.1 37 Intron XP_011544040.1
XM_011545739.1 37 Intron XP_011544041.1
Gene
UBFD1
Gene Name
ubiquitin family domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019116.2 37 UTR 5 NP_061989.2
XM_011545894.2 37 UTR 5 XP_011544196.1
XM_017023459.1 37 UTR 5 XP_016878948.1
XM_017023460.1 37 UTR 5 XP_016878949.1

View Full Product Details