Product Details

SNP ID
rs35629312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79361343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTACTTACCCAGCAAGAGGTCT[C/G]TGCGACGAAGGCGGAAGGACTTTCT
Phenotype
MIM: 300222
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITM2A PubMed Links

Gene Details

Gene
ITM2A
Gene Name
integral membrane protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171581.1 1026 Missense Mutation ACA,AGA T186R NP_001165052.1
NM_004867.4 1026 Missense Mutation ACA,AGA T230R NP_004858.1

View Full Product Details