Product Details
- SNP ID
-
hCV22273597
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58469949 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCACCCTCCTCCCCTCCCTCCCA[A/C]CCGATTCCCTTCCCTTCCATCATCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF324
PubMed Links
Gene Details
- Gene
- ZNF324
- Gene Name
- zinc finger protein 324
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014347.2 |
|
Intron |
|
|
NP_055162.1 |
- Gene
- ZNF446
- Gene Name
- zinc finger protein 446
There are no transcripts associated with this gene.
View Full Product Details