Product Details

SNP ID
rs2856997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32813999 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGTTGGCACCCATGGAGTTACC[A/C]CCCATCGGTTACAGATTCTCACAGC
Phenotype
MIM: 600629 MIM: 170261
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HLA-DOB PubMed Links

Gene Details

Gene
HLA-DOB
Gene Name
major histocompatibility complex, class II, DO beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002120.3 Intron NP_002111.1
Gene
TAP2
Gene Name
transporter 2, ATP binding cassette subfamily B member
There are no transcripts associated with this gene.

View Full Product Details