Product Details
- SNP ID
-
rs3741826
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:12814330 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCAAGTGAAATAAGGATATGGTCA[C/G]TAAAGGTACAAAGTTTGGGTAATGG
- Phenotype
-
MIM: 615428
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DDX47
PubMed Links
Gene Details
- Gene
- DDX47
- Gene Name
- DEAD-box helicase 47
View Full Product Details