Product Details
- SNP ID
-
rs1051374
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:12814193 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGGGATGGACAAAACCCACCAAGAT[C/T]CAGATTGAAGCTATTCCTTTGGCCT
- Phenotype
-
MIM: 615428
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDX47
PubMed Links
Gene Details
- Gene
- DDX47
- Gene Name
- DEAD-box helicase 47
View Full Product Details