Product Details

SNP ID
hCV22274165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45948865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCTATCCAGTATCTGGAGATC[C/T]TAAATGAAGAGGGAGGTGAGTCCTG
Phenotype
MIM: 610827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCIF1 PubMed Links

Gene Details

Gene
PCIF1
Gene Name
PDX1 C-terminal inhibiting factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022104.3 4240 Intron NP_071387.1
Gene
ZNF335
Gene Name
zinc finger protein 335
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022095.3 4240 UTR 3 NP_071378.1

View Full Product Details