Product Details

SNP ID
rs7523423
Assay Type
Validated
NCBI dbSNP Submissions
41
Location
Chr.1:234392627 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTTGGAGTAAATTAGAGTTAAAAAA[C/T]GTATTAACCTAAACTTAACTCTTTA
Phenotype
MIM: 614772 MIM: 605052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COA6 PubMed Links

Gene Details

Gene
COA6
Gene Name
cytochrome c oxidase assembly factor 6
There are no transcripts associated with this gene.

Gene
TARBP1
Gene Name
TAR (HIV-1) RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005646.3 Intron NP_005637.3
XM_005273234.3 Intron XP_005273291.1
XM_011544263.2 Intron XP_011542565.1
XM_017002194.1 Intron XP_016857683.1
XM_017002195.1 Intron XP_016857684.1
XM_017002196.1 Intron XP_016857685.1
XM_017002197.1 Intron XP_016857686.1

View Full Product Details