Product Details

SNP ID
rs35416065
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68645718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGACCTTGGAGGCGGGAGGCG[C/T]GGAGCAGGAGCCCGGCCAGGCGCTG
Phenotype
MIM: 114021
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CDH3 PubMed Links

Gene Details

Gene
CDH3
Gene Name
cadherin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317195.1 672 Missense Mutation GCG,GTG A43V NP_001304124.1
NM_001317196.1 672 Intron NP_001304125.1
NM_001793.5 672 Missense Mutation GCG,GTG A43V NP_001784.2
XM_011522800.2 672 Missense Mutation GCG,GTG A43V XP_011521102.1

View Full Product Details