Product Details

SNP ID
rs73032426
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:175324229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCCATTGTCTGCATTATTTTTC[C/T]CAAGACTCAAAATATTTCTTTTTAA
Phenotype
MIM: 601995
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNR PubMed Links

Gene Details

Gene
TNR
Gene Name
tenascin R
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003285.2 Intron NP_003276.3
XM_011509949.2 Intron XP_011508251.1
XM_017002218.1 Intron XP_016857707.1
XM_017002219.1 Intron XP_016857708.1

View Full Product Details