Product Details

SNP ID
rs11465472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49255621 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCCTCTTCCTTCCTTTATACCAG[T/C]CCTCGTCCCAGGTGAACTTGGTTTC
Phenotype
MIM: 300859 MIM: 300292
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CCDC22 PubMed Links

Gene Details

Gene
CCDC22
Gene Name
coiled-coil domain containing 22
There are no transcripts associated with this gene.

Gene
FOXP3
Gene Name
forkhead box P3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114377.1 Intron NP_001107849.1
NM_014009.3 Intron NP_054728.2
XM_006724533.2 Intron XP_006724596.2
XM_011543916.2 Intron XP_011542218.1
XM_017029565.1 Intron XP_016885054.1
XM_017029566.1 Intron XP_016885055.1
XM_017029567.1 Intron XP_016885056.1

View Full Product Details