Product Details
- SNP ID
-
hCV22275286
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:150325829 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGAGGCTGTGGAGGAAGGCCGAA[A/G]CTCTAGGCATTCCAAGTCTAGCAGT
- Phenotype
-
MIM: 607301
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRPF3
PubMed Links
Gene Details
- Gene
- PRPF3
- Gene Name
- pre-mRNA processing factor 3
There are no transcripts associated with this gene.
View Full Product Details