Product Details

SNP ID
rs11205325
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:150072257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTTAGTTCACTTTTTCAAACAT[A/G]TAACAACATCCCAGTTAGTGTGTTT
Phenotype
MIM: 610035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS45 PubMed Links

Gene Details

Gene
VPS45
Gene Name
vacuolar protein sorting 45 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001279353.1 Intron NP_001266282.1
NM_001279354.1 Intron NP_001266283.1
NM_001279355.1 Intron NP_001266284.1
NM_007259.4 Intron NP_009190.2
XM_017000164.1 Intron XP_016855653.1

View Full Product Details