Product Details

SNP ID
rs35926103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74750496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAAAAGCAAAAGAGAATGTAA[A/G]GAGGAAGAAAAGAAGAAAAACTATG
Phenotype
MIM: 600345
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTC PubMed Links

Gene Details

Gene
BTC
Gene Name
betacellulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316963.1 Intron NP_001303892.1
NM_001729.3 Intron NP_001720.1
XM_011532211.1 Intron XP_011530513.1
XM_017008568.1 Intron XP_016864057.1

View Full Product Details