Product Details
- SNP ID
-
rs2279112
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:119416784 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTGCCCTCCCCAGTGCCCGGGCC[A/G]GTGCCCTGCGACGCCGCCGACCTGC
- Phenotype
-
MIM: 164958
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NOV
PubMed Links
Gene Details
- Gene
- NOV
- Gene Name
- nephroblastoma overexpressed
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002514.3 |
346 |
Missense Mutation |
CAG,CGG |
Q42R |
NP_002505.1 |
View Full Product Details