Product Details

SNP ID
rs17167331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134158197 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTATACAGAAAGTTGCAGGTGCC[A/G]AAACTAGTAACTGTTAATACATTAA
Phenotype
MIM: 601434 MIM: 189908
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SKP1 PubMed Links

Gene Details

Gene
SKP1
Gene Name
S-phase kinase-associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006930.3 893 UTR 3 NP_008861.2
NM_170679.2 893 Intron NP_733779.1
Gene
TCF7
Gene Name
transcription factor 7 (T-cell specific, HMG-box)
There are no transcripts associated with this gene.

View Full Product Details