Product Details
- SNP ID
-
rs61735536
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:3752517 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACCAGGAAACTCACGCGGACAGC[A/G]GCCTGGCACGCAGCCAGGGGCAGCC
- Phenotype
-
MIM: 604262
MIM: 612689
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APBA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80066100] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APBA3
- Gene Name
- amyloid beta precursor protein binding family A member 3
- Gene
- TJP3
- Gene Name
- tight junction protein 3
There are no transcripts associated with this gene.
View Full Product Details