Product Details
- SNP ID
-
rs3731881
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219055690 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGACCTGGAAGGCTCTCAGCCTGTG[A/G]GGCTCGCGGTCCAGGAAAATGAGCA
- Phenotype
-
MIM: 600726
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IHH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76479554] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IHH
- Gene Name
- indian hedgehog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002181.3 |
802 |
Silent Mutation |
CCC,CCT |
P251P |
NP_002172.2 |
- Gene
- MIR3131
- Gene Name
- microRNA 3131
There are no transcripts associated with this gene.
View Full Product Details