Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001197301.1 | 393 | Intron | NP_001184230.1 | ||
NM_001197302.1 | 393 | Intron | NP_001184231.1 | ||
NM_014419.3 | 393 | Missense Mutation | AGG,ATG | R109M | NP_055234.1 |
XM_006723142.2 | 393 | Missense Mutation | AGG,ATG | R124M | XP_006723205.1 |
XM_011526724.1 | 393 | Missense Mutation | AGG,ATG | R132M | XP_011525026.1 |
XM_011526725.1 | 393 | Missense Mutation | AGG,ATG | R116M | XP_011525027.1 |
XM_011526726.2 | 393 | Missense Mutation | AGG,ATG | R116M | XP_011525028.1 |
XM_011526727.2 | 393 | Intron | XP_011525029.1 | ||
XM_011526728.2 | 393 | Intron | XP_011525030.1 | ||
XM_011526729.1 | 393 | Missense Mutation | AGG,ATG | R34M | XP_011525031.1 |
XM_017026593.1 | 393 | Missense Mutation | AGG,ATG | R34M | XP_016882082.1 |