Product Details

SNP ID
rs73790201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134158771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGACTTTTATGTGACCTTCTT[C/T]GTTATACCTCTATAATGTTGAATCC
Phenotype
MIM: 601434
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SKP1 PubMed Links

Gene Details

Gene
SKP1
Gene Name
S-phase kinase-associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006930.3 Intron NP_008861.2
NM_170679.2 Intron NP_733779.1

View Full Product Details