Product Details

SNP ID
rs2272002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53424132 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCCATGTGTCCCCATGGCAGGGC[A/T]CAGGTTCCAGGCCTCTGCTGACCCT
Phenotype
MIM: 600956 MIM: 189906
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AMHR2 PubMed Links

Gene Details

Gene
AMHR2
Gene Name
anti-Mullerian hormone receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164690.1 Intron NP_001158162.1
NM_001164691.1 Intron NP_001158163.1
NM_020547.2 Intron NP_065434.1
XM_011538173.1 Intron XP_011536475.1
XM_011538174.1 Intron XP_011536476.1
XM_011538176.1 Intron XP_011536478.1
XM_011538178.1 Intron XP_011536480.1
XM_011538179.1 Intron XP_011536481.1
XM_011538180.1 Intron XP_011536482.1
XM_011538181.1 Intron XP_011536483.1
XM_011538183.2 Intron XP_011536485.1
XM_011538184.2 Intron XP_011536486.1
XM_011538185.1 Intron XP_011536487.1
XM_011538186.2 Intron XP_011536488.1
XM_011538187.2 Intron XP_011536489.1
XM_017019179.1 Intron XP_016874668.1
Gene
SP1
Gene Name
Sp1 transcription factor
There are no transcripts associated with this gene.

View Full Product Details