Product Details
- SNP ID
-
rs3774396
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:42258245 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTCCAGAAGGAGCTACAAGGAGCAG[A/G]GAGGAAGGAAACAGGGACATTGCAT
- Phenotype
-
MIM: 118440
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCK
PubMed Links
Gene Details
- Gene
- CCK
- Gene Name
- cholecystokinin
View Full Product Details