Product Details

SNP ID
rs11569679
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.19:47871532 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
AATTTATCAAAGTCTTCAGCTTGGG[C/T]CACTGTGAAGTGATTTTTCCAGTCC
Phenotype
MIM: 125263
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01595 PubMed Links

Gene Details

Gene
LINC01595
Gene Name
long intergenic non-protein coding RNA 1595
There are no transcripts associated with this gene.

Gene
SULT2A1
Gene Name
sulfotransferase family 2A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003167.3 921 Missense Mutation ACC,GCC T261A NP_003158.2

View Full Product Details