Product Details

SNP ID
rs148647140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:148081528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACACAGACACAGGGGGCATGACA[C/T]GGGGGCCTCCTTTCCGACCCTGGAT
Phenotype
MIM: 609746 MIM: 600983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP10 PubMed Links

Gene Details

Gene
ARHGAP10
Gene Name
Rho GTPase activating protein 10
There are no transcripts associated with this gene.

Gene
NR3C2
Gene Name
nuclear receptor subfamily 3 group C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000901.4 Intron NP_000892.2
NM_001166104.1 Intron NP_001159576.1
XM_011531975.1 Intron XP_011530277.1
XM_011531976.2 Intron XP_011530278.1
XM_011531977.2 Intron XP_011530279.1
XM_011531978.2 Intron XP_011530280.1
XM_017008217.1 Intron XP_016863706.1

View Full Product Details