Product Details

SNP ID
rs148487929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72752253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACAGTTTAATCTGCTTTTATTT[A/C]TTTGGCTGTCTTCCAAACCACTTTC
Phenotype
MIM: 611861
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADPGK PubMed Links

Gene Details

Gene
ADPGK
Gene Name
ADP dependent glucokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031284.4 1617 UTR 3 NP_112574.3
XM_006720712.3 1617 UTR 3 XP_006720775.1
XM_006720713.3 1617 UTR 3 XP_006720776.1
XM_011522102.2 1617 UTR 3 XP_011520404.1
XM_011522103.2 1617 UTR 3 XP_011520405.1
XM_011522104.2 1617 Intron XP_011520406.1
XM_017022664.1 1617 UTR 3 XP_016878153.1

View Full Product Details