Product Details

SNP ID
rs17102350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:90913090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTATATGTTATAACAGTGTTTT[A/G]TGGTTTCCACCAGGAGTGATCTGCA
Phenotype
MIM: 609599 MIM: 606115
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD1 PubMed Links

Gene Details

Gene
ANKRD1
Gene Name
ankyrin repeat domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014391.2 Intron NP_055206.2
Gene
RPP30
Gene Name
ribonuclease P/MRP subunit p30
There are no transcripts associated with this gene.

View Full Product Details