Product Details

SNP ID
rs28377312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99386590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGAGGAGAGACTGAGTCATGAAA[A/G]GTGAGGTGTTGTTGCCTAGGTGCAC
Phenotype
MIM: 604223 MIM: 607075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPC1B PubMed Links

Gene Details

Gene
ARPC1B
Gene Name
actin related protein 2/3 complex subunit 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005720.3 Intron NP_005711.1
XM_006715825.1 Intron XP_006715888.1
XM_006715826.1 Intron XP_006715889.1
Gene
PDAP1
Gene Name
PDGFA associated protein 1
There are no transcripts associated with this gene.

View Full Product Details