Product Details

SNP ID
rs115757951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96006447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCTAGTTATTTTTGTTCAAGTA[C/T]AATTTAATAATACTTATTGGTTTAT
Phenotype
MIM: 151570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LTA4H PubMed Links

Gene Details

Gene
LTA4H
Gene Name
leukotriene A4 hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000895.2 Intron NP_000886.1
NM_001256643.1 Intron NP_001243572.1
NM_001256644.1 Intron NP_001243573.1
XM_005268871.1 Intron XP_005268928.1
XM_011538348.1 Intron XP_011536650.1
XM_011538349.2 Intron XP_011536651.1

View Full Product Details