Product Details

SNP ID
rs77321553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73445027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCTTACCATTGAGGCATATATT[A/G]TTTTTCCTGAAAAGCTTTATTAAAT
Phenotype
MIM: 114106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP3CB PubMed Links

Gene Details

Gene
PPP3CB
Gene Name
protein phosphatase 3 catalytic subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142353.2 Intron NP_001135825.1
NM_001142354.2 Intron NP_001135826.1
NM_001289968.1 Intron NP_001276897.1
NM_001289969.1 Intron NP_001276898.1
NM_021132.3 Intron NP_066955.1
XM_005269944.1 Intron XP_005270001.1
XM_017016379.1 Intron XP_016871868.1

View Full Product Details