Product Details

SNP ID
hCV25594036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54906175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCACTGCTCAGCACTAGGCCGG[C/T]AGAATCTGAGCGATGTCTTCCACAC
Phenotype
MIM: 604530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCR1 PubMed Links

Gene Details

Gene
NCR1
Gene Name
natural cytotoxicity triggering receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145457.2 26 UTR 5 NP_001138929.2
NM_001145458.2 26 UTR 5 NP_001138930.2
NM_001242356.2 26 UTR 5 NP_001229285.1
NM_001242357.2 26 UTR 5 NP_001229286.1
NM_004829.6 26 UTR 5 NP_004820.2

View Full Product Details