Product Details

SNP ID
rs3816716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84142638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATGTGGGTGGTACGTGTCTTT[C/G]TTTGCATGTGTGTCAGGAATATGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GHITM PubMed Links

Gene Details

Gene
GHITM
Gene Name
growth hormone inducible transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014394.2 Intron NP_055209.2

View Full Product Details